Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Universal Kinase Assay Kit, Fluorometric, Enzyme Activity, 250 Tests

Catalog No. 502583469
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3469 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3469 Supplier Abcam Supplier No. AB138879250TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay for measuring kinase activity based on the rate of ADP formation.

  • Assay Type: Enzyme activity
  • Detection Method: Fluorescent
  • Sample Types: Purified protein, Cell Lysate
  • Range: 1 to 300 μM
  • Sensitivity: < 0.3 μM
  • Applications: Compatible with fluorometric plate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Kinase
No. of Reactions 250 Tests
Detection Location In-Solution Detection
For Use With (Equipment) Fluorometric Plate Reader
Format Kit
Detection Method Fluorescence
Sample Type Purified Protein, Cell Lysate
Quantity 250 Tests
For Use With (Application) Kinase activity measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.