Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Universal Laboratory Hydrometer
SCI_ED

Catalog No. S80850 Shop All Eisco Products
Change view
Click to view available options
For Use With (Application):
Measuring Specific Gravity
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S80850 Measuring Specific Gravity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80850 Supplier Eisco™ Supplier No. PH0128
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Universal Laboratory Hydrometer measures specific gravity within the range of 0.700 to 2.000 with 0.01 graduations and a dual scale for light or heavy liquids.

  • Universal Laboratory Hydrometer
  • Includes hard plastic case
  • Measures specific gravity range 0.700 to 2.000
  • Dual Scale For Light and Heavy Liquids
  • Graduations: 0.01

Specifications

Type Hydrometer
Increments 0.01
Length (Metric) 387 mm
Includes Dual Scale
Length (English) 15.2 in.
For Use With (Application) Measuring Specific Gravity
Specific Gravity Range 0.700 to 2.000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.