Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Universal Lateral Flow Assay Kit, 100 Tests

Catalog No. 502656165
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-6165 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-6165 Supplier Abcam Supplier No. AB270537100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized kit designed for versatile lateral flow assay development.

  • Applications: LFA
  • Citation Count: 2

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Format Kit
No. of Reactions 100 Tests
Quantity 100 Tests
For Use With (Application) Lateral Flow Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.