Learn More
Eisco™ Universal Mirror
SCI_ED
Catalog No. S24359 Shop All Eisco Products
Change view
Click to view available options
Universal stainless-steel mirror
Catalog No.
S24359 Universal stainless-steel mirror
1 options
Catalog No. S24359 Supplier Eisco™ Supplier No. PH0528
Add to Cart
Add to Cart
Aids students in performing experiments using light rays with plane, concave and convex mirrors.
  • Made from stainless-steel
Specifications
Product Type Universal Mirror
Lens Material Stainless Steel
For Use With (Application) Study of optics
Includes One side flat and one side curved
Height (English) 0.75 in.
Length (English) 3.5 in.