Learn More
Labconco Universal PC Mount Kit
GSA_VASDP
Catalog No. 17200225 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Enclosures
Catalog No. For Use With (Equipment)
17-200-225 Enclosures
1 options
Catalog No. 17-200-225 Supplier Labconco Supplier No. 5315000
Add to Cart
Add to Cart
Universal PC Mount Kit provides a 4 x 7 in. area to secure most clamp style mounting systems for laptops, tablets and monitors. Mounts on either the side of the base stand. Compatible with all base stands.
  • Provides a 4 x 7 in. (10.2 x 17.8 cm) area to secure most clamp style mounting systems for laptops, tablets and monitors
  • Mounts on either the side of the Labconco base stands
  • Compatible with Labconco base stands
  • Articulating arms not included and sold separately
  • Installation required
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Enclosures
Product Type Universal PC Mount Kit