Learn More
Mascon Medical Mascon Medical™ Universal Pipette Tips, 10μl, 96 Tips/Rack, 10 Racks/Box, 5 Boxes/Case, Sterile, Filtered, Low-retention
SDP
Supplier:  Mascon Medical FSTIPS10F
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin, and Beckman automation systems. These high-precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features:•96 Tip/Nested Tip Rack Format•Meets ISO13485 quality management system standard•Nonpyrogenic•Low-retention•UniversalSpecifications:•Volume: 10µL•Qty. / Case: 4800•Size: 31.5mm•Tip Format: 96 Tips/Rack•Material: Polypropylene•Filter: Yes•Sterile: Yes•Packaging: Nested Tip Rack, 10 Racks/Pack, 5 Packs /Carton•Packaging Format: 50 Racks/Carton
Catalog No. NC2458380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.