Learn More
Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Catalog No. 13261406
Change view
Click to view available options
For Use With (Equipment):
1300 Series A2 BSC
3 ft. models w/SS interior, left
3 ft. models w/SS interior, right
3 ft. models, left and top/bottom
4 ft. models, left and top
4 ft. models, right and top
5 ft. models w/SS interior, left
5 ft. models w/SS interior, right
5 ft. models, left and top
5 ft. models, left and top/bottom
5 ft. models, right and top/bottom
6 ft. models, left and top
6 ft. models, right and top
all models, left and bottom
ft. models, right and top/bottom
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-261-406 6 ft. models, right and top
13-261-354 5 ft. models, right and top/bottom
13-261-352 ft. models, right and top/bottom
13-261-355 5 ft. models, left and top/bottom
13-261-353 3 ft. models, left and top/bottom
13-261-360 1300 Series A2 BSC
13-261-402 4 ft. models, right and top
13-261-403 4 ft. models, left and top
13-261-405 5 ft. models, left and top
13-261-407 6 ft. models, left and top
13-261-409 all models, left and bottom
13-261-357 3 ft. models w/SS interior, left
13-261-356 3 ft. models w/SS interior, right
13-261-358 5 ft. models w/SS interior, right
13-261-359 5 ft. models w/SS interior, left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 13-261-406 Supplier Thermo Scientific™ Supplier No. 1911421
Add to Cart
Add to Cart
Factory-installed Universal Air/Gas Piping
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Piping
For Use With (Equipment) 6 ft. models, right and top