Learn More
Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Description
Universal Piping for Air/Gas Service
- Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
- Two pre-plumbed service ports per side only
- Factory-installed; must be ordered with cabinet
- For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.
Specifications
Specifications
| Product Type | Universal Piping |
| For Use With (Equipment) | 6 ft. models, left and top |