Learn More
Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Description
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
- (1) microplate clamp
- (2) 25mL flask clamps
- (2) 50mL flask clamps
- (2) 125mL flask clamps
- (4) 250/300mL flask clamps
- (4) 500mL flask clamps
- (2) 1L flask clamps
- (2) 2L flask clamps
- Replacement screws
Note:
Replacement screws also available separately.
Specifications
Specifications
| Holds | Vials, Tubes |
| Product Type | Foam Insert |
| For Use With (Equipment) | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |