Learn More
Globe Scientific Universal Plug Stoppers
Catalog No. 22170555 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Light Blue
Catalog No. Color
22-170-555 Light Blue
1 options
Catalog No. 22-170-555 Supplier Globe Scientific Supplier No. 5529
Add to Cart
Add to Cart
Globe Scientific Inc. Universal Plug Stoppers produced from low density polyethylene (LDPE)
  • Fits most 10 mm, 12 mm, 13 mm, and 16 mm tubes
Specifications
Product Type Plug Stopper
Material LDPE
Color Light Blue
Sterility Non-sterile
Disposable Single-use