Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Universal Precaution Compliance Kit Refill

Catalog No. 19090911 Shop All Medique Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-911 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-911 Supplier Medique Supplier No. 17100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refill bag of the Universal Precaution Compliance Kit

  • Greatly reduce infectious risk factors your workers face every day from improper body-fluid spill clean up
  • This kit helps meet the OSHA Compliance 1910.1030 Bloodborne Pathogens Standard by combining personal protection and clean up items mandated by OSHA, CDC and State Health Departments
  • Combines PPE and clean-up items mandated by OSHA, CDC and State Health Departments
  • Includes: 
    • Pair of Vinyl Gloves
    • Protective Apron
    • Combo Mask/Safety Shield
    • Red Z™ Solidifier (10 g Pouch)
    • Scoop/Scraper
    • SaniZide Pro™ Germicidal Wipe (Dimensions: 8 x 8 in.)
    • Red Bio-hazard Waste Bag (Dimensions: 24 x 24 in.)
    • Twist Tie
    • p.a.w.s.™ Antimicrobial Hand Wipe (Dimensions: 5 x 8 in.)
    • Identification Tag and Instructions

Specifications

Product Type Universal Precaution
For Use With (Application) Reduce exposure to infectious waste and cross-contamination
Length (Metric) 5 in.
Color White
Material Plastic
Sterility Sterile
Quantity 1 Ea.
Size Standard
Width (Metric) 8 in.
Height (Metric) 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.