Learn More
Medique Universal Precaution Kit with Plastic Case
Catalog No. 19090610 Shop All Medique Products
Change view
Click to view available options
For Use With (Application):
Cleaning, Transport, Disposal Of 1L Infectious Blood, Body Fluid Spills
Catalog No. For Use With (Application)
19-090-610 Cleaning, Transport, Disposal Of 1L Infectious Blood, Body Fluid Spills
1 options
This item is not returnable. View return policy
Catalog No. 19-090-610 Supplier Medique Supplier No. SS17102
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
1 Red Z™ Solidifier, 1 Biozide™ hard surface disenfectant, 1 PAWS™ antimicrobial hand wipe, 1 pair Vinyl gloves, 1 Scoop/scraper, 1 Combination mask/face shield, 1 Apron, 1 Biohazard bag, 1 Identification tag, 1 Instruction card
To aid in the clean-up, transportation and disposal of 1L of infectious blood or body fluid spills.
  • Kit includes Red Z Solidifier, Biozide hard surface disinfectant, PAWS antimicrobial hand wipe, Vinyl gloves, Scoop/scraper, Combination mask/face shield, Apron, Biohazard bag, Identification tag, Instruction card
Specifications
For Use With (Application) Cleaning, Transport, Disposal Of 1L Infectious Blood, Body Fluid Spills
Product Type Universal Precaution Kit
Includes 1 Red Z™ Solidifier, 1 Biozide™ hard surface disinfectant, 1 PAWS™ antimicrobial hand wipe, 1 pair Vinyl gloves, 1 Scoop/scraper, 1 Combination mask/face shield, 1 Apron, 1 Biohazard bag, 1 Identification tag, 1 Instruction card