Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Universal Rat Reference RNA for QuantiGene Assays

Catalog No. QS0641
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0641 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QS0641 Supplier Invitrogen™ Supplier No. QS0641
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Universal Rat Reference RNA is an RNA extract used as a positive control in QuantiGene assays.

Universal Rat Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Rat Reference RNA is composed of total RNA from 14 rat cell lines:

  • Blood, basophilic leukemia
  • Blood, T-lymphocyte, lymphoma
  • Blood, B-lymphoblast, hybridoma
  • Brain, glioma
  • Embryo, yolk sac carcinoma
  • Embryo, fibroblast, normal
  • Kidney, normal
  • Liver, hepatoma
  • Lung, alveolar macrophage, normal
  • Lung, alveolar type II, normal
  • Mammary gland, adenocarcinoma
  • Muscle, myoblast
  • Skin, normal
  • Testis, Leydig cell tumor

Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.

TRUSTED_SUSTAINABILITY

Specifications

Species Rat
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Reference RNA
Quantity 10 μg
Sample Source Cell Line
Shipping Condition Shipped at 2°C to 8°C

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.