Description
Includes
CHO cell-expressed Recombinant Mouse IMPAD1, PAP, Phosphatase Buffer 3, Phosphate Standard, Malachite Green Reagents A and B
Uses inositol monophosphatase 3 (IMPAD1) as a coupling phosphatase to remove inorganic phosphate quantitatively from the leaving nucleotide 3-phosphoadenosine-5-phosphate (PAP) that is generated during sulfotransferase reactions
No time-consuming separat
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |