Learn More
PBL Assay Science Universal Type I Interferon Alpha (IFN-Alpha, IFNa) [1e6 (1,000,000) units/vial]
Supplier:  PBL Assay Science 112002
Encompass_Preferred
Recombinant protein hybrid Universal Type I Interferon is manufactured by PBL Assay Science (Catalog #11200-2) for research use only (RUO) and packaged in 1e6 (1,000,000) units/vial. This cytokine exhibits activity across multiple species and is essential to in vivo or in vitro studies where the species-relevant Type I IFN-Alpha is unavailable or performs inconsistently.
Catalog No. 50-153-7964
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.