Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Universal Type I IFN Protein
GSA_VA

Catalog No. 112001
Change view
Click to view available options
Quantity:
1 MU
100000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
112001 100000 U
112002 1 MU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 112001 Supplier R&D Systems™ Supplier No. 112001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

IgG Polyclonal antibody specifically detects IgG in Human samples. It is validated for Control.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA)
Purification Method Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography
Quantity 100000 U
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.