Learn More
R&D Systems™ Universal Type I IFN Protein
Description
Specifications
Specifications
| Formulation | Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) |
| Purification Method | Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography |
| Quantity | 1 MU |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Gene Alias | IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.