Learn More
Cayman Chemical Senae folIum SenIdIn A 5mg
SDP
Supplier:  Cayman Chemical 352375 MG
Encompass_Preferred
A STAT3 inhibitor (IC50 0 74 UM in a reporter assay) and a derivative of WP1066 inhibits proliferation of HeLa Caco-2 A549 A375 U87MG and HL-60 cancer cells (IC50s 1 8 1 8 3 2 8 2 3 and 1 2 UM respectively) active against H pylori and C jejuni (MICs 1 6 and 4 7 UM respectively)
Catalog No. 50-291-3460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.