Learn More
Honeywell™ North™ 17 mil Butyl Gloves
Catalog No. 19085590 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-590 X-Small
19-085-591 Small
19-085-593 Medium
19-085-594 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-085-590 Supplier Honeywell™ Supplier No. B174RGI/BAT/XS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Butyl gloves with Grip-Saf™ palm feature high permeation resistance to gas and water vapor, and is also highly resistant to ketones and esters.
  • For greater worker protection, especially when handling toxic substances
  • Flexible and sensitive, even at lower temperatures
  • Curved finger and hand design provides a better fit for enhanced comfort
Specifications
Certifications/Compliance ASTM D3389-05, ASTM F790-97, EN 374
Certifications/Compliance (Europe) EN 420
Color Black
Latex Free Yes
Material Butyl
Size X-Small
Thickness (English) 17 mil
Thickness (Metric) 0.43 mm
EN388 Standard EN 388
Glove Cuff Style Beaded
Grip Design Grip-Saf™
Lining Unlined
Model B174RGI/BAT/XS
Length (English) 14 in.
Length (Metric) 35.5 cm