Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ 17 mil Butyl Gloves

Catalog No. 19085594 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-594 Large
19-085-590 X-Small
19-085-591 Small
19-085-593 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-085-594 Supplier Honeywell™ Supplier No. B174RGI/BAT/L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Butyl gloves with Grip-Saf™ palm feature high permeation resistance to gas and water vapor, and is also highly resistant to ketones and esters.

  • For greater worker protection, especially when handling toxic substances
  • Flexible and sensitive, even at lower temperatures
  • Curved finger and hand design provides a better fit for enhanced comfort

Specifications

Certifications/Compliance ASTM D3389-05, ASTM F790-97, EN 374
Certifications/Compliance (Europe) EN 420
Applications Industrial
Color Black
Latex Free Yes
Material Butyl
Product Type Butyl Gloves
Size Large
Thickness (English) 17 mil
Thickness (Metric) 0.43 mm
EN388 Standard EN 388
Glove Cuff Style Beaded
Grip Design Grip-Saf™
Lining Unlined
Model B174RGI/BAT/L
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.