Learn More
Description
Specifications
Specifications
| Formulation | 20 mM Tris-phosphate (pH 7.5), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15 % (v/v) Glycerol. |
| For Use With (Application) | 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring |
| Quantity | 500 μL |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
