Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UoS 12258
GREENER_CHOICESDP

Catalog No. 635350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-535-0 50 mg
63-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-535-0 Supplier Tocris Bioscience™ Supplier No. 6353/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Positive allosteric modulator of AMPA receptors (pEC50 = 5.2 for potentiation of AMPA-mediated currents in vitro)

UoS 12258 is a positive allosteric modulator of AMPA receptors (pEC50 = 5.2 for potentiation of AMPA-mediated currents in vitro). Also potentiates AMPA-mediated synaptic responses in vivo. Improves performance of rats in various learning and memory paradigms

Specifications

Chemical Name or Material N-[(2S)-5-(6-Fluoro-3-pyridinyl)-2,3-dihydro-1H-inden-2-yl]-2-propanesulfonamide
CAS 875927-64-3
Quantity 50 mg
Molecular Formula C17H19FN2O2S
Formula Weight 334.41
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.