Learn More
TARGETMOL CHEMICALS INC UPACICALCET 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T29067
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Upacicalcet (AJT-240) is an intraventricular calcium mimetic a SHPT available for human hemodialysis that inhibits excessive parathyroid hormone (PTH) secretion by directly acting on parathyroid cell membrane calcium-sensitive receptors thereby lowering blood levels of PTH.Upacicalcet is an orthosteric modulator of calcium-sensitive receptors and prevents adenine-induced secondary vascular calcification and bone disease in a rat model of adenine-induced secondary hyperparathyroidism. purity: 98%
Catalog No. 50-258-0469
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.