Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Upadacitinib
GREENER_CHOICESDP

Catalog No. 778350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-835-0 Supplier Tocris Bioscience™ Supplier No. 7783/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

JAK1-selective inhibitor

Upadacitinib is a potent and selective JAK1 inhibitor (IC50 = 47 nM). In a cellular assay, Upadacitinib shows selectivity for JAK1 over JAK2, JAK3 and TYK2 (IC50 values are 0.014 μM, 0.593 μM, 1860 μM and 2715 μM). In vivo, Upadacitinib suppresses paw swelling and bone destruction in a rat model of arthritis.

Specifications

Chemical Name or Material (3 S,4 R)-3-Ethyl-4-(3 H-imidazo[1,2-a]pyrrolo[2,3-e]pyrazin-8-yl)-N-(2,2,2-trifluoroethyl)-1-pyrrolidinecarboxamide
CAS 1310726-60-3
Quantity 50 mg
Target Jak1
Molecular Formula C17H19F3N6O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.