Learn More
MilliporeSigma™ Supelco™ Upchurch Fingertight Fittings Kit
Catalog No. 111009945 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Upchurch Fingertight Fittings Kit
Catalog No.
11-100-9945 Upchurch Fingertight Fittings Kit
1 options
Catalog No. 11-100-9945 Supplier MilliporeSigma™ Supelco™ Supplier No. 58632
Add to Cart
Add to Cart
High pressure fittings with 10-32 threads for 1/16 in. O.D. tubing.
Kit includes: Delrin Fingertight nut, good to 4,000 psi (280 kg/cm2), 5 ea. Stainless steel Fingertight nut, good to 6,000 psi (420 kg/cm2), wrenchtight to 10,000 psi (700 kg/cm2), 5 ea. PEEK gripping ferrule, 20 ea.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications