Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Upchurch Microtight™ Tubing Sleeves

Catalog No. p-7191390 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Green
Diameter (Metric) Inner:
380 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Diameter (Metric) Inner
11-101-1850 Green 380 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-1850 Supplier MilliporeSigma™ Supelco™ Supplier No. 502626
Only null left
Add to Cart
Add to Cart

PEEK sleeves with all Microtight™ fittings can be used to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Diameter (English) Inner 0.015 in.
Diameter (Metric) Inner 380 μm
Packaging Quantity 10/Pk.

Microtight is a registered trademark of IDEX Health & Science, LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.