Learn More
MilliporeSigma™ Supelco™ Upchurch PEEK Fingertight Fitting
Catalog No. 111020897 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Nut and ferrule 1/4-28
Nut and ferrule 10-32
Packaging Quantity:
20/Pk.
30/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Packaging Quantity
11-102-0897 Nut and ferrule 10-32 30/Pk.
11-100-4749 Nut and ferrule 1/4-28 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0897 Supplier MilliporeSigma™ Supelco™ Supplier No. 57654
Add to Cart
Add to Cart
For use with 1/16 in. O.D. tubing and can be used to 6,000 psi (420 kg/cm2).
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 30/Pk.
Description Nut and ferrule 10-32
Material PEEK