Learn More
MilliporeSigma™ Supelco™ Upchurch PEEK LiteTouch™ Fitting, PEEK
Catalog No. 111005094 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Double-winged nut
Nut
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-5094 Double-winged nut
11-100-5093 Nut
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5094 Supplier MilliporeSigma™ Supelco™ Supplier No. 57653
Add to Cart
Add to Cart
Unique design prevents twisting of PEEK tubing.
The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers' stainless steel nuts. Order nuts and ferrules separately.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Packaging Quantity 10/Pk.
Description Double-winged nut
Material PEEK
LiteTouch is a registered trademark of IDEX Health & Science, LLC