Learn More
MilliporeSigma™ Supelco™ Upchurch Polymer Tubing Cutter
Catalog No. 111020947 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Upchurch Polymer Tubing Cutter
Catalog No.
11-102-0947 Upchurch Polymer Tubing Cutter
1 options
Catalog No. 11-102-0947 Supplier MilliporeSigma™ Supelco™ Supplier No. 57665U
Add to Cart
Add to Cart
Upchurch designed this reliable and durable device specifically for cutting 1/8 in. - 1/16 in. PEEK, PTFE, and Tefzel tubing. Includes 4 replacement blades.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications