Learn More
Idex Bulkhead Union
Catalog No. 05700329
Click to view available options
Diameter (English) Inner:
0.04 in.
Catalog No. Diameter (English) Inner
05-700-329 0.04 in.
1 options
Catalog No. 05-700-329 Supplier Idex Supplier No. P441
Thread through the equipment housing to connect the external system to tubing
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom port
  • 2.9 μL swept volume
  • Assembly includes SS nut and washer
Specifications
Diameter (English) Inner 0.04 in.