Learn More
Idex Threaded Plugs and Caps
Catalog No. 05701443
Click to view available options
Type:
Plug
Color:
Natural
Material:
PEEK
Port Type:
5/16-24 Flat Bottom
Catalog No. Type Color Material Port Type
05-701-443 Plug Natural PEEK 5/16-24 Flat Bottom
1 options
This item is not returnable. View return policy
Catalog No. 05-701-443 Supplier Idex Supplier No. P556
This item is not returnable. View return policy
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
Type Plug
Color Natural
Port Type 5/16-24 Flat Bottom
Material PEEK