Learn More
Idex Tubing Expanding Awl
Catalog No. 0570190
Click to view available options
For Use With (Equipment):
Tubing Used with Barbed Fittings and Also Works with Semi-Rigid Tubing
Catalog No. For Use With (Equipment)
05-701-90 Tubing Used with Barbed Fittings and Also Works with Semi-Rigid Tubing
1 options
Catalog No. 05-701-90 Supplier Idex Supplier No. P780
Expands tubing for barbed fittings
  • Stainless steel
  • For use with semi-rigid tubing such as FEP and PFA
Specifications
Type Tubing Expanding Awl
Material Stainless Steel
For Use With (Equipment) Tubing Used with Barbed Fittings and Also Works with Semi-Rigid Tubing