Learn More
MilliporeSigma™ Supelco™ Upchurch Tubing Adapter, 1/4-28 Female, PEEK
Catalog No. 111005244 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Female Luer, 1/4-28 Female
Male Luer, 1/4-28 female
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-5244 Male Luer, 1/4-28 female
11-100-5256 Female Luer, 1/4-28 Female
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5244 Supplier MilliporeSigma™ Supelco™ Supplier No. 55072
Add to Cart
Add to Cart
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It can be used to connect capillary tubes in HPLC, GC.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Diameter (English) Outer 1/4-28 female
Diameter (English) Inner male Luer (lock hub-quick connect)
Packaging Quantity 1/Pk.
Description Male Luer, 1/4-28 female
Material PEEK