Learn More
ATCC UPCI:SCC154; Squamous Cell Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3241
Encompass
UPCI:SCC154 is a squamous cell line that was isolated in 1996 from the tongue of a 54-year-old, White male patient with carcinoma. The cells form islands and the monolayer does not become confluent. These cells are positive for Human Papilloma Virus (HPV) which is of interest to many researchers because HPV is a major etiologic factor for oropharyngeal squamous cell carcinoma (OPSCC). These cells have no TP53 mutations as assayed by sequencing the 5-8 exons of TP53 and showed no amplification of chromosomal band 11q13 using FISH. This cell line was deposited by S Gollin and can be used for studying the initiation, cancerization, prognosis, intervention, and treatment of oral cancers.
Catalog No. 50-238-1812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.