Learn More
Selleck Chemical LLC UPF 1069 10mg 1048371-03-4
SDP
Supplier:  Selleck Chemical LLC S803810MG
Encompass_Preferred
UPF 1069 is a selective PARP2 inhibitor with IC50 of 0.3 ?M. It is ~27-fold selective against PARP1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5594
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.