Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Freezer Rack, (1) Drawers for Storage Bottles w/H<4 3/16" (106mm), Rack Dimensions 22 3/16L x 4 1/2H x 5 1/2W" (564 x 114 x 140mm), Drawer Area 20 5/8 x 4 1/2" (523 x 114mm)
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFDLB101
Encompass_Preferred
Rack Designed to store bottles < 4 3/16 (106mm)H, in 1 Drawer, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack. Great for organizing -20⁰, -30⁰ , -86⁰ Freezers and Cold Rooms
Catalog No. NC2208090
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.