Learn More
Thomas Scientific Upright Freezer Rack, Stainless Steel, 9 1/4"H x 22 1/8"D x 5 1/2"W,Holds 16 Std 2 Inch Boxes
SDP
Supplier:  Thomas Scientific 1149C29
Encompass
In addition to cryogenic freezer equipment, we supply upright freezer racks, chest freezer racks, liquid nitrogen freezer racks, canisters / cassettes and frames as well as laboratory boxes and dividers. By understanding the needs of the biotechnology industry, Custom Biogenic Systems provides superior laboratory cryogenic freezers and complete inventory control rack systems that support veterinarian, horticulture, pharmaceutical, industrial, scientific and research laboratories and banks.
Catalog No. 50-195-996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.