Learn More
Enterprise Technology Solutions CyberPower PR3000RTXL2UHVACN Smart App Sinewave UPS (3000W)
Supplier: Enterprise Technology Solutions PR3000RTXL2UHVACN
The 2000VA and 2200W PR3000RTXL2UHVACN Smart App Sinewave UPS from CyberPower offers robust power protection and management in a rackmount/tower convertible form factor. This unit provides pure sinewave output, ensuring clean, stable power for up to 6 devices, providing ample time in the event of a power failure for proper equipment shutdown.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.