Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate In Vivo Shape Reagent (Live Cell RNA Structure Analysis)

Catalog No. 03310mi Shop All MilliporeSigma Products
Change view
Click to view available options
Product Type:
In Vivo Shape Reagent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
03-310-MI In Vivo Shape Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-310-MI Supplier MilliporeSigma™ Supplier No. 03310
Only null left
Add to Cart
Add to Cart

Ready to use RNase-free solution with low toxicity to use with live cells for determination of RNA structure

  • High cell-permeability for rapid modification of RNA
  • Reacts with all RNA nucleotides for high resolution mapping
  • Contains highly pure form of 2-methylnicotinic acid imidazolide (NAI)
  • Forms 2'-O-adducts on single stranded regions of RNA that can be detected by Selective 2'-Hydroxyl Acylation Primer Extension (SHAPE)
  • NAI rapidly absorbed into cells, formulated to have low levels of toxicity for use with live cells
  • Reacts in unbiased fashion with all four RNA nucleotides
  • Combination of low toxicity, rapid uptake lack of nucleotide bias enables high resolution RNA structure analysis in variety of living systems
  • Packaging: Vial
  • Color: Clear, Colourless

Recommended Storage

Store NAI at −20°C in a non-frost-ree freezer protected from light

TRUSTED_SUSTAINABILITY

Specifications

Description 2-methylnicotinic acid imidazolide (NAI) in DMSO In Vivo Shape Reagent
Product Type In Vivo Shape Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.