Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRFS1 Monoclonal Antibody (OTI4H8)
GREENER_CHOICE

Catalog No. PIMA527471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527471 Supplier Invitrogen™ Supplier No. MA527471
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UQCRFS1 Monoclonal Antibody for Western Blot, IHC (P)

Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRFS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Uqcrfs1
Gene Accession No. P20788, P47985, Q9CR68
Gene Alias 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1
Gene Symbols Uqcrfs1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 79-274 of UQCRFS1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291103, 66694, 7386
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.