Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uracil, MP Biomedical

Catalog No. MP210320491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210320491 1 kg
MP210320480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210320491 Supplier MP Biomedicals, Inc Supplier No. 0210320491
Add to Cart
Add to Cart

Uracil HPLC 99.9% is a pyrimidine derivative.

  • Used in drug delivery and in pharmaceuticals
  • Used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes
  • Uracil is produced by hydrolysis of nucleic acid and used in biochemical research
  • It serves as an allosteric regulator and coenzyme for reactions in the human body and in plants
  • It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, pyrimidine-2,4 1h,3h-dione, 2,4-dioxopyrimidine, 2,4-pyrimidinedione, pirod, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Absorbance UV/Vis: l max (water) 258 ± 2 nm; OD250/OD260 0.82-0.88; OD280/OD260 0.0-0.2
Melting Point >300°C (literature)
Color White
pH Neutral to Litmus Paper (4% aq susp)
Packaging Plastic
Quantity 1 kg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.