Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uranium AA Standard, 1000 ppm U in 3% HNO3, Ricca Chemical
SDP

Catalog No. AU1KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AU1KN-100 100 mL Natural Poly Bottle
AU1KN-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AU1KN-100 Supplier Ricca Chemical Company Supplier No. AU1KN100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1344-58-7 , 7697-37-2 , 7732-18-5
Molecular Formula O3U
Molecular Weight (g/mol) 286.026
InChI Key JCMLRUNDSXARRW-UHFFFAOYSA-N
PubChem CID 74013
IUPAC Name trioxouranium
SMILES O=[U](=O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 4.76, 0.11
CAS Max % 97.0, 4.95, 0.12
Physical Form Liquid
Color Yellow
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg U (1, 000ppm U)UO3 in 3% HNO3
Chemical Name or Material Uranium AA Standard
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.