Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uranyl Acetate, Dihydrate, Reagent, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607644 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-644 5 g
18-607-645 25 g
18-607-646 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-644 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. U10065GM
Add to Cart
Add to Cart

U1006, 6159-44-0, C4H6O6U·2H2O

Spectrum™ Chemical Uranyl Acetate, Dihydrate, Reagent , also known as Uranyl ethanoate, is used as a negative stain in electron microscopy. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6159-44-0
Molecular Formula C4H12O8U
Molecular Weight (g/mol) 426.16
InChI Key COQLPRJCUIATTQ-UHFFFAOYSA-N
IUPAC Name uranyl acetate
SMILES O.O.O=[U]=O.CC(O)=O.CC(O)=O

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 98 to 102%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.