Learn More
Electron Microscopy Sciences Uranyl Formate 1 GR

Supplier: Electron Microscopy Sciences 22450
CAS #16984-59-1 UO₂ (CHO₂) ₂.H₂O F.W. 378.08
Melting point: 110°C
For ultra fine grain structure, our Uranyl Formate offers amazing advancement in negative staining for microscopy.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.