Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URB1/NPA1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569829
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9829 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9829 Supplier Bethyl Laboratories, Inc Supplier No. A305215AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The German cDNA consortium; Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases. Slavov D., Gene 247:215-232(2000). Nagase T., DNA Res. 5:31-39(1998).

Specifications

Antigen URB1/NPA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene URB1
Gene Accession No. O60287
Gene Alias C21orf108, KIAA0539, NOP254, NPA1
Gene Symbols URB1
Host Species Rabbit
Immunogen Between 2221 and 2271
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9875
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.