Learn More
Sigma Aldrich Fine Chemicals Biosciences Endothelial cell growth su5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E275915MG
Encompass
Endothelial cell growth supplement (ECGS) comprises acidic fibroblast growth factor (afGf) and basic fGf (bfGf).
Catalog No. 50-290-0423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.