Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607654 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-654 500 g Poly Bottle
18-607-655 2.5 kg Poly Pail
18-607-656 12 kg Poly Pail
18-607-657 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-654 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. U1018500GM
Add to Cart
Add to Cart

U1018, 57-13-6, CH4N2O

Spectrum™ Chemical Urea, FCC is a component of animal feed, providing a relatively cheap source of nitrogen to promote growth. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.0 to 100.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.