Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, For Analysis, ACS, MilliporeSigma™

Catalog No. M1084875000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1084875000 500 g Plastic Bottle
M1084871000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1084875000 Supplier MilliporeSigma Supplier No. 1084875000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 133°C
Density 1.34g/3 (20°C)
pH 9 (100g/L, H2O, 20°C)
Packaging Plastic Bottle
Quantity 500 g
Solubility Information 1000g/L
Vapor Pressure <0.1 hPa (20°C)
Formula Weight 60.06
Percent Purity 99.0 to 100.5%
Grade ACS/ISO
EINECSNumber 200-315-5
RTECSNumber YR6250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.