Learn More
National Diagnostics UreaGel Loading Buffer | 10 x 1mL
SDP
Supplier:  National Diagnostics EC857
Encompass_Preferred
UreaGel Loading Buffer is a denaturing loading buffer for UreaGel and other denaturing polyacrylamide gel applications. UreaGel Loading Buffer contains 95% formamide, 18mM EDTA, SDS, xylene cyanole and bromophenol blue.
Catalog No. NC0217043
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.