Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Urea Nitrogen (BUN) Colorimetric Detection Kit

Catalog No. EIABUN
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIABUN 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIABUN Supplier Invitrogen™ Supplier No. EIABUN
Add to Cart
Edge
Add to Cart

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Urea Nitrogen (BUN) levels in Serum, Plasma, Urine, Saliva and Tissue Culture Media.

The Urea Nitrogen (BUN) Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of urea nitrogen in serum, plasma, urine, saliva and tissue culture media samples.

This complete, ready-to-use kit includes clear 96-well plate(s), urea nitrogen standard, and other components to perform the assay. A 96-well microplate reader capable of reading optical absorption at 450 nm is required for use of this kit.

Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: serum, plasma, urine, saliva, and tissue culture media samples
• Sensitivity: 0.030 mg/dL
• Standard curve range: 0.156–10 mg/dL
• Reactivity: species independent

Background
Urea is a by-product of protein metabolism by the liver, and is therefore removed from the blood by the kidneys. Urea freely filters through the glomerulous, but is reabsorbed by the renal tubules in a flow-dependent fashion. The higher the flow rate, the greater amount of urea nitrogen is cleared from circulation and eliminated through the kidneys. Urea nitrogen is identical across all species and this kit will measure urea nitrogen from sources other than human.

Assay principle
The Urea Nitrogen ((also called BUN) Colorimetric Detection Kit is designed to quantitatively measure urea nitrogen in a variety of samples. A urea nitrogen standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Samples are mixed with color reagents A and B and incubated at room temperature for 30 minutes. The colored product is read at 450 nm.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at 2–8°C.
Immunoassay Kit Format Colorimetric Detection Kit
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 2 x 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Urea Nitrogen (BUN)
Is the Urea Nitrogen (BUN) Colorimetric Detection Kit (Cat. No. EIABUN) based on an antibody-antigen reaction or chemical reaction?

The Urea Nitrogen (BUN) Colorimetric Detection Kit (Cat. No. EIABUN) is based on a chemical reaction, in which urea reacts with Reagent A to form a Schiff’s base, which then reacts with Reagent B to form a colored product.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.